Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stearic Acid, Powder (Triple Pressed), N.F., J.T.Baker™

Click to view available options
Quantity:
500g
1.5kg
Packaging:
Poly Bottle
57-11-4
C18H36O2
284.484
QIQXTHQIDYTFRH-UHFFFAOYSA-N
stearic acid,stearophanic acid,n-octadecanoic acid,cetylacetic acid,pearl stearic,stearex beads,octadecansaeure,stearinsaeure,vanicol,1-heptadecanecarboxylic acid
5281
CHEBI:28842
octadecanoic acid
CCCCCCCCCCCCCCCCCC(=O)O
57-11-4
C18H36O2
284.484
QIQXTHQIDYTFRH-UHFFFAOYSA-N
stearic acid,stearophanic acid,n-octadecanoic acid,cetylacetic acid,pearl stearic,stearex beads,octadecansaeure,stearinsaeure,vanicol,1-heptadecanecarboxylic acid
5281
CHEBI:28842
octadecanoic acid
CCCCCCCCCCCCCCCCCC(=O)O
Specific Gravity 1L = 0.94kg
Packaging Poly Bottle
Quantity 1.5kg
Flash Point 196.00°C
Formula Weight 284.48
Physical Form Powder (Triple Pressed)
Grade NF
Chemical Name or Material Stearic Acid