Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stearic Acid (Prill/Laboratory), Fisher Chemical

Catalog No. a292500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g

Chemical Identifiers

CAS 57-11-4
Molecular Formula C18H36O2
Molecular Weight (g/mol) 284.48
MDL Number MFCD00002752
InChI Key QIQXTHQIDYTFRH-UHFFFAOYSA-N
Synonym stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid
PubChem CID 5281
ChEBI CHEBI:28842
IUPAC Name octadecanoic acid
SMILES CCCCCCCCCCCCCCCCCC(O)=O

Specifications

Melting Point 67°C
Color White
Density 0.840g/cm³
Boiling Point 361°C
Flash Point 196°C
Packaging Poly Bottle
Linear Formula CH3(CH2)16COOH
Quantity 500 g
Vapor Pressure 1mmHg at 174°C
Formula Weight 284.49
Grade Laboratory
Physical Form Solid
Chemical Name or Material Stearic Acid, Prill
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.