Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stearoyl Chloride 97.0+%, TCI America™
SDP

Catalog No. S0404250G
Click to view available options
Quantity:
25 g
250 g

Chemical Identifiers

CAS 112-76-5
Molecular Formula C18H35ClO
Molecular Weight (g/mol) 302.927
MDL Number MFCD00000744
InChI Key WTBAHSZERDXKKZ-UHFFFAOYSA-N
PubChem CID 8212
IUPAC Name octadecanoyl chloride
SMILES CCCCCCCCCCCCCCCCCC(=O)Cl

Specifications

Color Yellow
Boiling Point 215°C
Quantity 250 g
UN Number 3265
Formula Weight 302.93
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Stearoyl Chloride
EINECSNumber (2)-0632&(2)-3552
RTECSNumber RG2162000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.