Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stearyl Stearate Test Solution, Restek™
SDP

Catalog No. 06802086 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-802-086 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-802-086 Supplier Restek Corporation Supplier No. 31636
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2778-96-3 , 110-82-7
Molecular Formula C36H72O2
Molecular Weight (g/mol) 536.97
InChI Key NKBWPOSQERPBFI-UHFFFAOYSA-N
PubChem CID 17720
IUPAC Name octadecyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCCCC

Specifications

CAS Min % 0.2
Ignition Point 245°C
CAS Max % 99.800000
Odor Pungent
Flash Point -4°C
Specific Gravity 0.779 to 0.784 g/cm3 at 20°C
Concentration 2,000 μg/mL in Cyclohexane
UN Number UN1145
Color Colorless
Solubility Negligible
Type Single-component Standard
Quantity 10 mL
Chemical Name or Material Stearyl stearate
Show More Show Less
Hazard Category Danger
Hazard Statement Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Very toxic to aquatic life.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilation and lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wash hands and skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection
ShelfLife 6 months
EINECSNumber 203-806-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.