Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Public Seating™ Steel Height Adjustable Science Lab Table with HPL Top

Click to view available options
Dimensions (D x W x H):
48 x 24 x 22.25 to 37.25 in.
54 x 24 x 22.25 to 37.25 in.
60 x 24 x 22.25 to 37.25 in.
60 x 30 x 22.25 to 37.25 in.
72 x 24 x 22.25 to 37.25 in.
72 x 30 x 22.25 to 37.25 in.
Description
- 16 gauge steel frame apron and 14 gauge legs
- 11 gauge steel tabletop support tabs and corner brackets fuse as one with four heavy-duty tamper-free Loctite hex screws
- Adjustable height steel legs feature a supreme dual connection
- Leg-base is fastened to tabletop with four heavy-duty screws which gets inserted into the frame using a strong welded hangar bolt
- Legs connect to frame corner brackets with industrial-grade nylon-insert lock nuts
- Legs adjust in 1 in. increments and lock firmly in place with two K-9 external tooth lock washers and heavy duty tamper-free hex screws
- All legs come standard with simple-adjust glides with optional casters
- High Pressure Laminate table top provides the ideal surface for lab experiments
Specifications
Specifications
Product Type | Science Lab Table |
Certifications/Compliance | MAS2 |
For Use With (Application) | Industrial Use |
Top Style | HPL |
Warranty | 10 years |
Color | Black |
Dimensions (D x W x H) | 54 x 24 x 22.25 to 37.25 in. |
Height (English) | 22.25 to 37.25 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction