Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stella/DPPA3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982100
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2100 Supplier Bioss Supplier No. BS12280R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DPPa3 (stella-related protein) is a stem cell marker protein which is restricted to pluripotent human embryonic stem cells, premeiotic germ lineage cells and testicular germ cell tumor cells. Otherwise known as ′developmental pluripotency-associated protein 3,′ it is expressed in primordial germ cells, Oocytes, preimplantation embryos and pluripotent cells. This protein has a SAP-like domain and a splicing factor motif-like structure, suggesting possible roles in chromosomal organization or RNA processing.

Specifications

Antigen Stella/DPPA3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DPPA3
Gene Accession No. Q6W0C5
Gene Alias 2410075G02Rik; Cap1p; Compaction-associated protein 1; Crg1; developmental pluripotency associated 3; developmental pluripotency-associated 3; Developmental pluripotency-associated protein 3; DPPA3; PCG7; Pgc7; primordial germ cell 7; Primordial germ cell protein 7; Stella; STELLAR; Stella-related protein
Gene Symbols DPPA3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Stella/DPPA3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 359787
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.