Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stella/Dppa3 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$148.20 - $378.00
Specifications
Antigen | Stella/Dppa3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Stella/Dppa3 Polyclonal antibody specifically detects Stella/Dppa3 in Mouse, Rat samples. It is validated for Western BlotSpecifications
Stella/Dppa3 | |
Polyclonal | |
Purified | |
RUO | |
Mouse, Rat | |
developmental pluripotency associated 3, DPPA3, DPPA3 developmental pluripotency associated 3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human Stella/Dppa3 (NP_631964.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Developmental Biology | |
PBS, 50% glycerol, pH7.3 | |
359787 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title