Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Stella Monoclonal Antibody (283910)
GREENER_CHOICE

Catalog No. PIMA523986
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523986 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523986 Supplier Invitrogen™ Supplier No. MA523986
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

DPPa3 (stella-related protein) is a stem cell marker protein which is restricted to pluripotent human embryonic stem cells, premeiotic germ lineage cells and testicular germ cell tumor cells. Otherwise known as 'developmental pluripotency-associated protein 3,' it is expressed in primordial germ cells, Oocytes, preimplantation embryos and pluripotent cells. This protein has a SAP-like domain and a splicing factor motif-like structure, suggesting possible roles in chromosomal organization or RNA processing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Stella
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 283910
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene DPPA3
Gene Accession No. Q8QZY3
Gene Alias 2410075G02Rik; Cap1p; Compaction-associated protein 1; Crg1; developmental pluripotency associated 3; developmental pluripotency-associated 3; developmental pluripotency-associated protein 3; Dppa3; PCG7; Pgc7; primordial germ cell 7; primordial germ cell protein 7; Stella; STELLAR; Stella-related protein
Gene Symbols DPPA3
Host Species Rat
Immunogen E. coli-derived recombinant mouse Stella/Dppa3 Met1-Asn150.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 73708
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.