Learn More
Invitrogen™ Stella Polyclonal Antibody

Description
The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_b) and 94% homologous in chimp.
Specifications
Specifications
| Antigen | Stella |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide |
| Gene | DPPA3 |
| Gene Accession No. | Q6W0C5 |
| Gene Alias | 2410075G02Rik; Cap1p; Compaction-associated protein 1; Crg1; developmental pluripotency associated 3; developmental pluripotency-associated 3; developmental pluripotency-associated protein 3; Dppa3; PCG7; Pgc7; primordial germ cell 7; primordial germ cell protein 7; Stella; STELLAR; Stella-related protein |
| Gene Symbols | DPPA3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.