Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Stella Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115972
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115972 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115972 Supplier Invitrogen™ Supplier No. PA5115972
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Stella.

DPPa3 (stella-related protein) is a stem cell marker protein which is restricted to pluripotent human embryonic stem cells, premeiotic germ lineage cells and testicular germ cell tumor cells. Otherwise known as 'developmental pluripotency-associated protein 3,' it is expressed in primordial germ cells, Oocytes, preimplantation embryos and pluripotent cells. This protein has a SAP-like domain and a splicing factor motif-like structure, suggesting possible roles in chromosomal organization or RNA processing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Stella
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene DPPA3
Gene Accession No. Q6W0C5, Q8QZY3
Gene Alias 2410075G02Rik; Cap1p; Compaction-associated protein 1; Crg1; developmental pluripotency associated 3; developmental pluripotency-associated 3; developmental pluripotency-associated protein 3; Dppa3; PCG7; Pgc7; primordial germ cell 7; primordial germ cell protein 7; Stella; STELLAR; Stella-related protein
Gene Symbols DPPA3
Host Species Rabbit
Immunogen A synthesized peptide derived from human DPPA3(Accession Q6W0C5), corresponding to amino acid residues A97-A147.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 359787, 73708
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.