Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Stem Cell Qualified ECM Gel
Shop All MilliporeSigma Products
Click to view available options
Quantity:
5 mL
10 x 5 mL
Description
- Stem Cell Qualified ECM Gel is a soluble basement membrane extract (BME) purified from Engelbreth-Holm-Swarm (EHS) tumor that has been pre-qualified for human ES/iPS cell culture
- The matrix polymerizes at 20 to 40°C to form a reconstituted basement membrane
- ECM Gel contains laminin, collagen IV, entactin and heparan sulfate among other ECMs
- The matrix has been extensively tested and optimized for human ES/iPS cell culture
- Eliminates the need for time-consuming prescreening and lot identification
- Provides an optimized ECM coating necessary for long term culture of pluripotent human ES/iPS cells
- Supports feeder-free culture of human ES/iPS cells in serum-free media
- Compatible with all feeder free human ES/iPS cell expansion media (i.e. mTeSR™, PluriSTEM™)
Specifications
Specifications
| Form | Gel |
| For Use With (Application) | Human ES and iPS cell culture |
| Quantity | 5 mL |
| Assays | < 5% spontaneous differentiation for 3 passages using Human iPS cells |
| Endotoxin Level | < 20 EU/mL |
| Color | Red, clear of particulates |
| Content And Storage | Store at -20°C, thaw overnight on ice in a 2 to 8°C refrigerator |
| Description | Basement membrane extract (BME) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction