Learn More
K'NEX Education™ STEM Explorations: Swing Ride Building Set

Description
Requires
2 AA batteries
Using the materials included in this set, middle-school aged children will be engaged and energized as they further their knowledge and understanding of the science, technology, engineering and math concepts associated with a real-life amusement park ride
Once built, a downloadable booklet guides students through 3 hands-on, inquiry-based experiments on each of the models. Students will learn about how mass affects speed, how to gear-up and gear down, and much more.
Set includes:
- More than 470 parts plus a battery-powered motor for a single child or team of 2-3 children to build a working Swing Ride
- Two additional models: a Ferris Wheel and a Boom Ride (each model can be built one at a time)
The Experiment Guide is aligned to national STEM standards and is appropriate for grade levels 5 to 9.
Specifications
Specifications
Certifications/Compliance | Experiment Guide is aligned to national STEM standards |
Age | 8 |
For Use With (Application) | Teaching STEM concepts |
Includes | 470 parts, battery-powered motor, 2 additional models: Ferris Wheel and a Boom Ride |
Requires | 2 “AA” (LR6) Batteries not included |
Class Size | Team of 2 to 3 Students |
Product Type | Swing ride building set |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.