Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Learning Resources STEM Explorers™ Geomakers

Click to view available options
Class Size:
1 Student
Description
- With the help of the included challenges, kids craft their own 2-D and 3-D shapes during STEM-filled play sessions that boost early geometry, critical thinking, and fine motor skills.
Specifications
Specifications
| Grade Level | Elementary, Intermediate - Middle School, High School |
| Grade | K to 12 |
| Class Size | 1 Student |
| Product Type | STEAM Learning |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction