Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hand2Mind STEM Lab Makerspace Cart

Click to view available options
Class Size:
1 Class
Description
- This cart provides plenty of space for you to create a centralized resource in a STEM lab or library.
Specifications
Specifications
| Grade Level | Elementary, Intermediate - Middle School, High School |
| Grade | K to 12 |
| Class Size | 1 Class |
| Product Type | Classroom Makerspace Cart |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction