Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ StemXVivo EMT Inducing Media Supplement (100X)
SDP

Catalog No. CCM017 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CCM017 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. CCM017 Supplier R&D Systems™ Supplier No. CCM017

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

To reliably induce epithelial to mesenchymal transition (EMT) in vitro

Reliable induction of EMT is useful for understanding the biology of EMT and for identifying and evaluating novel prognostic markers and therapeutics for fibrotic diseases and metastasis. However, the ability to induce EMT in vitro typically requires TGF-beta stimulation or labor-intensive genetic modification. Moreover, although TGF-beta is necessary to induce EMT, it is not sufficient in all cell types.

  • Drives EMT in 5 days to ensure efficient use of time and reagents
  • Provides a straightforward method to induce EMT in vitro
  • Is completely defined to reduce experimental variability
  • May induce EMT in cells resistant to TGF-beta stimulation alone
  • Avoids time-consuming genetic modifications

Specifications

Product Type Stem Cell Supplement Media
Product Line R&D Systems
Quantity 1 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.