Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    STEP Antibody, mFluor Violet 500 SE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281893MFV500
Description
STEP Polyclonal antibody specifically detects STEP in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| STEP | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| EC 3.1.3.48, FLJ14427, Neural-specific protein-tyrosine phosphatase, protein tyrosine phosphatase, non-receptor type 5 (striatum-enriched), protein-tyrosine phosphatase striatum-enriched, STEPPTPSTEP, Striatum-enriched protein-tyrosine phosphatase, tyrosine-protein phosphatase non-receptor type 5 | |
| STEP antibody was raised against an 18 amino acid peptide near the amino terminus of human STEP. The immunogen is located within amino acids 180 - 230 of STEP. Amino Acid Squence: LRTPPEPPTPLPPEDRRQ | |
| 0.1 mL | |
| Alzheimers Research, Cancer, Cellular Markers, Hypoxia, Neurodegeneration, Neuroscience, Protein Phosphatase, Signal Transduction | |
| 84867 | |
| Store at 4C in the dark. | |
| IgG | 
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            