Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Step by Step Number Line
SCI_ED

Catalog No. S79389
Click to view available options
Grade:
Pre-K to 12

Catalog No. S79389

Supplier: Learning Resources, Inc LER 0420

Only null left
Add to Cart
Add to Cart

Walk-on mat develops gross motor coordination.

Physically reinforces fundamentals in number recognition, sequencing and counting from 0-20. Made of durable vinyl. Dimensions: 1 x 20 ft. (0.3 x 6m).

Order Info

Grades PK+

Specifications

Grade Level Early Childhood, Elementary, Intermediate - Middle School, High School
Grade Pre-K to 12
Format Hands-on Learning
Product Type Step-by-step Number Line
Includes Sequencing and counting from 0 to 20
For Use With (Application) Physically reinforces fundamentals in number recognition
Subject Mathematics
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.