Learn More
Invitrogen™ STEP Polyclonal Antibody

Description
Immunogen sequence: EHTPIIVMIT NIEEMNEKCT EYWPEEQVAY DGVEITVQKV IHTEDYRLRL ISLKSGTEER GLKHY Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 91%.
Specifications
Specifications
| Antigen | STEP |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | PTPN5 |
| Gene Accession No. | P54829 |
| Gene Alias | FLJ14427; Neural-specific protein-tyrosine phosphatase; protein tyrosine phosphatase, non-receptor type 5; protein tyrosine phosphatase, non-receptor type 5 (striatum-enriched); protein-tyrosine phosphatase striatum-enriched; protein-tyrosine-phosphatase non-receptor 5; PTN5; PTP STEP; PTPN 5; PTPN5; PTPSTEP; STEP; STEP61; striatal-enriched protein tyrosine phosphatase; striatum-enriched protein-tyrosine phosphatase; Tyrosine-protein phosphatase non-receptor type 5 |
| Gene Symbols | PTPN5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.