Learn More
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Non-sterile
Catalog No. 07201479 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
0.05 mL
0.5 mL
1.25 mL
2.5 mL
5 mL
12.5 mL
25 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
07-201-479 5 mL
07-201-477 1.25 mL
07-201-478 2.5 mL
07-201-476 0.5 mL
07-201-481 25 mL
07-201-475 0.05 mL
07-201-480 12.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 07-201-479 Supplier Corning™ Supplier No. 6624
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Non-sterile provide precise, positive-displacement dispensing of viscous or volatile liquids.
  • Ideal for viscous, dense and volatile liquides
  • Minimum increments printed on the syringe itself
  • Accurately deliver selected dose
  • Made from high quality polypropylene and polyethylene
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Certifications/Compliance ISO8655
Material Polypropylene
Sterility Non-sterile
For Use With (Application) Dispensing of viscous and volatile liquids
Quantity 100/Pk.
Volume (Metric) 5 mL
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov