Learn More
Stoelting™ Stereotaxic Robot Drill and Microinjector Robot
SDP
Catalog No. 10000192 Shop All Stoelting Co Products
Click to view available options
Animal:
Mouse
Rat
Stoelting Stereotaxic Robot Drill and Microinjector Robot provides for a fully software driven solution to stereotaxic surgery
  • Allows for high precision injection.
  • After loading the syringe, the injection robot allows for multiple injections in different animals without tool exchange or recalibration
  • Completely automated solution for stereotaxic surgery
  • Features:
    • High Throughput Drill and Inject
    • No Tool Exchange
    • Define/Store Protocols
    • Precise Depth Control
    • Brain Windowing
    • Contour Drilling
    • Smooth Edges
    • Skull Thinning
Specifications
For Use With (Application) Neuroscience
Animal Rat
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.