Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Sterigmatocystin Monoclonal Antibody (4D3)

Catalog No. PIMA549620
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549620 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549620 Supplier Invitrogen™ Supplier No. MA549620

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Sterigmatocystin is a toxic secondary metabolite produced by certain species of fungi, including Aspergillus versicolor and Aspergillus nidulans. It is closely related to aflatoxin, another fungal toxin, and has been shown to have carcinogenic and mutagenic properties. Sterigmatocystin is commonly found in contaminated grains and nuts, and can also be present in feed for livestock.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sterigmatocystin
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 4D3
Concentration 7.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias Sterigmatocystine
Host Species Mouse
Immunogen Conjugate of STG hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.