Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Air-Tite™ Sterile Hypodermic Needles

Catalog No. 14817169 Shop All Air-Tite Products
Click to view available options
Needle Gauge:
14 G
16 G
18 G
20 G
21 G
22 G
23 G
25 G
26 G
27 G
Length (English) Needle:
0.25 in.
0.375 in.
0.50 in.
0.625 in.
0.75 in.
1 in.
1.5 in.
2 in.
3.25 in.
3.5 in.
4 in.
Certifications/Compliance:
ISO 13485
ISO 9001 Certified
Color:
Black
Blue
Brown
Gray
Green
Orange
Pink
White
Yellow

Air-Tite™ Sterile Hypodermic Needles are high quality, yet economical for laboratory and veterinary use only

  • Disposable Needles
  • Sterile
  • Polypropylene Hub
  • Stainless Steel Cannula
  • Single-use
  • Latex Free
  • ISO
  • Lab/Vet

Specifications

Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001 Certified
Material Polypropylene (Hub), Stainless Steel (Cannula)
Color Orange
Needle Gauge 25 G
For Use With (Application) Laboratory / Veterinary
Latex Free Yes
Length (English) Needle 1.5 in.
Product Type Leur
Disposable Single-use
Quantity 100/Box
Packaging Individually Wrapped
Needle Point Style Tri-Bevel
Connection Type Luer Lock
Graduated No
Sterility Sterile
Show More Show Less

Not for human use - laboratory and veterinary use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.