Learn More
StatLab Sterile Instrument Packs
Click to view available options
Includes:
Forceps, Scalpel, Scissors
Forceps, Scalpel, Scissors, Cutting Board
Description
- Cost effective and disposable, autoclaved Sterile Instrument Packs are designed to minimize contamination
- Ideal for possible infectious material
- Set contains: 1 forceps stainless steel 11mm, 1 scalpel with rigid plastic holder, No. 11 blade, 1 scissor stainless steel pointed
Choose between:
- 12 Sets/Pk.
- 10 Sets/Pk with Cutting Board included
Specifications
Specifications
Includes | Forceps, Scalpel, Scissors |
Quantity | 12 Sets/Pk. |
Product Type | Standard Dissecting Kit |