Learn More
StatLab Sterile Instrument Packs
SDP
Cost-effective pre-sterilized stainless steel instrument packs. Optional cutting board.
$193.70 - $288.50
Specifications
Product Type Standard Dissecting Kit
View More Specs
Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
22-130-490
SDP
View Documents
StatLab
903
Forceps, Scalpel, Scissors
Pack of 12 for $193.70
Only null left
Add to Cart
22-130-491
SDP
View Documents
StatLab
906
Forceps, Scalpel, Scissors, Cutting Board
Pack of 10 for $288.50
Only null left
Add to Cart
Description
Description
  • Cost effective and disposable, autoclaved Sterile Instrument Packs are designed to minimize contamination
  • Ideal for possible infectious material
  • Set contains: 1 forceps stainless steel 11mm, 1 scalpel with rigid plastic holder, No. 11 blade, 1 scissor stainless steel pointed
Choose between:
  • 12 Sets/Pk.
  • 10 Sets/Pk with Cutting Board included
Specifications
Specifications
Standard Dissecting Kit
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.