Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Sterile Syringe Filter PES

Catalog No. 09720514 Shop All Fisherbrand Products
Change view
Click to view available options
Pore Size:
0.22 μm
0.45 μm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
09-720-514 0.45 μm
09-720-511 0.22 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-720-514
Add to Cart
Add to Cart

Used for clarifying solutions.

Sterile filtration of protein solutions, tissue culture media, additives, buffers, and water

Individually wrapped
TRUSTED_SUSTAINABILITY

Specifications

Material (Membrane) PES
Pore Size 0.45 μm
Sterility Sterile
Housing Material Acrylic
Inlet Style Female Luer Lock
Outlet Style Male Leur-Slip
Quantity 50/Pk.
Diameter (Metric) 33 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.