Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, USP Purified/nSTERILE Filtered, WFI Quality, Ricca Chemical
SDP

Catalog No. R919200020S Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
20 L
200 L
Packaging:
Fiberboard Cube
Poly Drum
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

pH ∼7
Color Clear, Colorless
Melting Point 0°C
CAS Min % 100
Boiling Point 100°C
CAS Max % 100
Conductivity ≤1.3 μS/cm at 25°C
Description WFI Quality
Endotoxin ≤0.25 EU/mL
UN Number 2853.90.9090
Percent Purity 100%
Grade USP
Packaging Fiberboard Cube
Sterility Sterile
Density 1.00 g/cm3
Certificate of Analysis Yes
Quantity 20 L
Specific Gravity 1
Physical Form Liquid
Assay No assay needed
Chemical Name or Material Water
Show More Show Less
ShelfLife 12 Months
Recommended Storage Ambient/Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.