Learn More
Stoelting™ Sterilizing Pouches
SDP
Catalog No. p-7163818 Shop All Stoelting Co Products
Click to view available options
Dimensions (L x W):
12 x 15 in.
3.5 x 5.25 in.
3.5 x 9 in.
5.25 x 10 in.
7.25 x 13 in.
Includes:
Box of 200
Box of 400
Stoelting™ Sterilizing Pouches are compatible with steam, chemical vapor and EO Gas sterilizing processes. Self-sealing.
Specifications
For Use With (Application) Neuroscience
Dimensions (L x W) 3.5 x 5.25 in.
Includes Box of 200
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.