Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STERIS Coverage™ Spray TB Ready-to-Use Disinfectant
Click to view available options
Volume (English):
22 oz.
Description
Ideal for use in laboratories, hospitals, doctors' offices, nursing homes, schools, and veterinary clinics.
- Effective against Mycobacterium tuberculosis (BCG) and other bacteria, the HIV-1 virus, and pathogenic fungi in the presence of 5% organic soil (5% blood serum)
- Deodorizes areas that are difficult to keep fresh smelling, such as garbage storage areas and restrooms
- Recommended for use on metal, stainless steel, plastic, porcelain, glazed ceramic tile, and other nonporous environmental surfaces
Specifications
Specifications
Form | Liquid |
pH | 6.5 to 8.7 |
Appearance | Clear |
Certifications/Compliance | OSHA |
Composition | 21% Isopropanol |
For Use With (Application) | Used on floors, walls, Metal surfaces, Stainless Steel surfaces, Plastic surfaces, Porcelain, glazed Ceramic tile, bathrooms, shower stalls, bathtubs and other nonporous environmental surfaces |
Volume (English) | 22 oz. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction