Learn More
STERIS Lotion Soft™ Skin Conditioner Skin Cream
Catalog No. p-2916620
Click to view available options
Volume (Metric):
115 mL
444 mL
998.1 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
Requires
14-415-30 requires use of Steris Dispensing System (14-415-21)
Moisturizes skin without compromising the antimicrobial activity of products containing chlorhexidine gluconate (CHG)
  • Does not contain petrolatum or mineral oil—suitable for use under gloves
  • Contains emollients and other softening ingredients that make it ideal for use after antimicrobial handwashing
Specifications
Volume (Metric) 444 mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.