Learn More
STERIS Lotion Soft™ Skin Conditioner Skin Cream
Shop All STERIS Corporation Products
Click to view available options
Volume (Metric):
115 mL
444 mL
998.1 mL
Description
Requires
14-415-30 requires use of Steris Dispensing System (14-415-21)
Moisturizes skin without compromising the antimicrobial activity of products containing chlorhexidine gluconate (CHG)
- Does not contain petrolatum or mineral oil—suitable for use under gloves
- Contains emollients and other softening ingredients that make it ideal for use after antimicrobial handwashing
Specifications
Specifications
Product Type | Skin Conditioning Lotion |
Volume (Metric) | 998.1 mL |