Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Steritop™ E-GP Funnel-less Sterile Vacuum Filtration Systems
GREENER_CHOICE

Catalog No. p-8889003 Shop All MilliporeSigma Products
Click to view available options
Outlet Fitting:
38 mm threaded
45 mm threaded

The newest addition to the Stericup™ Quick Release Family. Evolved with an eco-conscience.

Stericup E and Steritop E sterile vacuum systems are the newest addition to the Stericup Quick Release Family. The Stericup E and Steritop E family of devices are designed as a more sustainable option for sterile filtration that significantly reduces plastic and packaging. These are funnel-less filter products for use in the sterile vacuum filtration of aqueous solutions such as media and tissue culture reagents/buffers.

The Stericup E and Steritop E devices are designed specifically for sterile filtration from compatible bottles (38 and 45 mm neck size) of media and tissue culture reagents/buffers.

SEGPT0038 is a 38 mm threaded collar, which uses a 0.22 μm pore size fast flow PES Express PLUS membrane that can directly connect to any volume, 45 mm receiver flask (glass or plastic) for sterilizing tissue culture media and buffers.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear with blue adapter
Disposable Single-use
Material Polystyrene
Material (Membrane) PES Express PLUS
Pore Size 0.22 μm
Sterility Sterile
Filtration Area 40 cm²
For Use With (Application) Sterile filtration
Outlet Fitting 38 mm threaded
Outlet Style 38 mm threaded
Type Bottle Top Filter
Volume (Metric) <2 mL (limited by vial volume)
Diameter (Metric) 73 mm
Diameter (Metric) Filter 73 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.