Learn More
United Scientific™ SteriTrue™ Sterile Sampling Scoops, Polystyrene
Description
United Scientific™ SteriTrue™ PS sterile sampling scoops are unique designs developed with end-users! The polystyrene scoops are ergonomically designed for long periods of use, they can hold stated volumes, and each scoop has a flat bottom surface for stability during weighing. The most unique feature of the scoops is the long handle: the cross ribs add structural support to help prevent breakage, and the bottom curved design fits comfortably in the palm of the hand.
- Ergonomically designed with voice-of-customer input
- Flat bottom surface for stability during weighing
- Gamma sterilized, individually packaged
- Lot stamped for traceability
- PS material meets FDA guidelines and is safe for food, pharma, and other uses
- Available in Pack and Case quantities
Specifications
Specifications
Product Type | Scoop |
Material | Polystyrene |
Autoclavable | Not Autoclavable |
Capacity (Metric) | 60 mL |
Quantity | 10/Pk. |
Cancer and Reproductive Harm
Your input is important to us. Please complete this form to provide feedback related to the content on this product.