Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sterol carrier protein 2 Antibody (OTI1E4), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274401B
Description
Sterol carrier protein 2 Monoclonal specifically detects Sterol carrier protein 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Sterol carrier protein 2 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
SCP2 | |
Full length recombinant protein of human Sterol carrier protein 2 (NP_002970) produced in HEK293T cells. | |
0.1 mL | |
6342.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1E4 | |
Western Blot, Immunohistochemistry | |
DKFZp686C12188, DKFZp686D11188, NLTP, non-specific lipid-transfer protein, NSL-TP, Propanoyl-CoA C-acyltransferase, SCP-2, SCP-CHI, SCPX, SCP-X, sterol carrier protein 2EC 2.3.1.176, Sterol carrier protein X | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction