Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STF 118804

Catalog No. 52075
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-075 5 mg
52-072-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-075 Supplier R&D Systems Supplier No. 5207/5
This item is not returnable. View return policy

NAMPT Inhibitors

STF 118804 is a NAMPT inhibitor (nicotinamide phosphoribosyltransferase, visfatin, PBEF1); inhibits NAD+ synthesis from nicotinamide. Potently inhibits the viability of multiple B-cell acute lymphoplastic leukemia (B-ALL) cell lines (IC50 < 10 nM); induces apoptosis in MF-411 cells. Induces regression of ALL xenografts and eliminates leukemia stems cells from bone marrow in mice.

Specifications

Chemical Name or Material 4-[5-Methyl-4-[[(4-methylphenyl)sulfonyl]methyl]-2-oxazolyl]-N-(3-pyridinylmethyl)benzamide
CAS 894187-61-2
Assay Percent Range >98%
Quantity 5 mg
Molecular Formula C25H23N3O4S
Formula Weight Observed MW: 461.53
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.