Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ STF 31
GREENER_CHOICE

Catalog No. 448410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-841-0 10 mg
44-845-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-841-0 Supplier Tocris Bioscience™ Supplier No. 4484/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

NAMPT inhibitor; also GLUT1 inhibitor

Inhibitor of GLUT1; inhibits glucose uptake in renal cell carcinoma (RCC) 4 cells. Activity causes necrotic cell death in von Hippel-Lindau (VHL)-deficient RCC cells. Also NAMPT inhibitor. Eliminates human pluripotent stem cells from culture with limited toxicity towards differentiated cells.

Chemical Identifiers

CAS 724741-75-7
Molecular Formula C23H25N3O3S

Specifications

Quantity 10 mg
Formula Weight 423.53
Purity >99%
Molecular Weight (g/mol) 423.531
Chemical Name or Material STF 31
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.