Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stigmasterol, 95%, Thermo Scientific Chemicals

Catalog No. ac132760050
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-48-7
Molecular Formula C29H48O
Molecular Weight (g/mol) 412.70
MDL Number MFCD00003630
InChI Key HCXVJBMSMIARIN-PHZDYDNGSA-N
Synonym stigmasterol, stigmasterin, beta-stigmasterol, stigmasta-5,22-dien-3beta-ol, 24s-5,22-stigmastadien-3beta-ol, unii-99wuk5d0y8, stigmasta-5,22e-dien-3beta-ol, stigmasta-5,22-dien-3-beta-ol, 3beta,22e-stigmasta-5,22-dien-3-ol, stigmasta-5,22-dien-3-ol, 3b,22e
PubChem CID 5280794
ChEBI CHEBI:28824
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-17-[(E,2R,5S)-5-ethyl-6-methylhept-3-en-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC[C@H](\C=C\[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)C

Specifications

Melting Point 161.0°C to 170.0°C
Color White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 8944
Specific Rotation Condition - 50.00 (20.00°C c=2,CHCl3)
Specific Rotation − 50.00
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 412.7
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Stigmasterol, 95%
Show More Show Less
EINECSNumber 201-482-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.