Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STIL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565446
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565446 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565446 Supplier Invitrogen™ Supplier No. PA565446
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLLEAQSLMPC SPKTTAVEDT VQAGRQMELV SVEAQSSPGL HMRKGVSIAV STGASLFWNA AGEDQEPDSQ MKQDDTKI Highest antigen sequence identity to the following orthologs - mouse 85%, rat 81%.

Plays an essential role in early embryonic development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STIL
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STIL
Gene Accession No. Q15468
Gene Alias cassiopeia; chunp6907; csp; MCPH7; nbb; scl/tal1 interrupting locus; SCL-interrupting locus protein; SCL-interrupting locus protein homolog; SIL; sill; Stil; STIL centriolar assembly protein; stil protein; TAL1 (SCL) interrupting locus like; Tal1 interrupting locus; TAL1/SCL interrupting locus-like protein; TAL-1-interrupting locus protein; zgc:110502
Gene Symbols STIL
Host Species Rabbit
Immunogen Recombinant Human STIL. Recombinant protein control fragment (Product #RP-104823).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6491
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.