Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STIM1 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11060547AF700
Description
STIM1 Polyclonal specifically detects STIM1 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.Specifications
STIM1 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated | |
Q13586 | |
STIM1 | |
STIM1 antibody was made to a synthetic peptide from the C-terminal region (within residues 600-685) of the human STIM1 protein. [UniProt# Q13586] | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine, Canine | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GOKD11S4896E, stromal interaction molecule 1 | |
Rabbit | |
77 kDa | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Plasma Membrane Markers, Signal Transduction | |
6786 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction