Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STIM1 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11060547V
Description
STIM1 Polyclonal specifically detects STIM1 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.Specifications
STIM1 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated | |
Q13586 | |
STIM1 | |
STIM1 antibody was made to a synthetic peptide from the C-terminal region (within residues 600-685) of the human STIM1 protein. [UniProt# Q13586] | |
Affinity Purified | |
RUO | |
Primary | |
Human, mouse, rat, bovine and canine. Immunogen has 93% homology to porcine and 79% homology to Xenopus. | |
Store at 4C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GOKD11S4896E, stromal interaction molecule 1 | |
Rabbit | |
77 kDa | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Plasma Membrane Markers, Signal Transduction | |
6786 | |
Human, Mouse, Rat, Bovine, Canine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction