Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STIM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517145
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517145 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517145 Supplier Thermo Scientific Supplier No. PA517145
Add to Cart
Add to Cart

STIM1 Polyclonal Antibody

STIM1 Polyclonal Antibody for Western Blot, IP

TRUSTED_SUSTAINABILITY

Specifications

Antigen STIM1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 19 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene STIM1
Gene Accession No. P70302, P84903, Q13586
Gene Alias D11S4896E; GOK; IMD10; RGD:1306831}; SIM; Stim1; stim1 {ECO:0000312; STIM1L; STRMK; Stromal interaction molecule 1; stromal interaction molecule 1 precursor; TAM; TAM1
Gene Symbols STIM1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the sequence of human STIM1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20866, 361618, 6786
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.