Learn More
Invitrogen™ STIM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582412
Description
Immunogen sequence: TAKQALSEVT AALRERLHRW QQIEILCGFQ IVNNPGIHSL VAALNIDPSW MGSTRPNPAH FIMTDDVDDM DEEIVSPLSM QSPSLQSSVR QRLTEPQHGL GSQRDLTHSD SESSLHMSDR QRVAPKPPQM SRAADEALNA MTSNGSH Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 98%.
STIM1 (stromal interacting molecule; also known as GOK) acts as a sensor of calcium depletion within the endoplasmic reticulum and transduces the signal to Orai1, the presumptive CRAC channel at the plasma membrane. Following decrease of luminal calcium concentration, STIM1 oligomerizes and induces Orai1 to enable entry of extracellular calcium into the cytoplasm. However, the precise mechanism of STIM1-Orai1 interaction has not been elucidated yet. Many questions also remain to be solved around STIM1 functional distribution. It turns out that STIM1 associates with growing ends of microtubules and is involved in endoplasmic reticulum tubule extension.
Specifications
| STIM1 | |
| Polyclonal | |
| Unconjugated | |
| STIM1 | |
| D11S4896E; GOK; IMD10; RGD:1306831}; SIM; Stim1; stim1 {ECO:0000312; STIM1L; STRMK; Stromal interaction molecule 1; stromal interaction molecule 1 precursor; TAM; TAM1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6786 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13586 | |
| STIM1 | |
| Recombinant protein corresponding to Human STIM1. Recombinant protein control fragment (Product #RP-90417). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.