Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STIM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596668
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596668 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596668 Supplier Invitrogen™ Supplier No. PA596668
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RQRVAPKPPQ MSRAADEALN AMTSNGSHRL IEGVHPGSLV EKLPDSPALA KKALLALNHG LDKAHSLMEL SPSAPPGGSP HLDSSRSHSP SSPDPDTPSP VGDSRALQAS RNTRIPHLAG KKAVAEEDNG SIGEETDSSP GRKKFPLKIF KKPLKK; Positive Samples: LO2, Jurkat, BxPC-3, HeLa, Mouse brain, Mouse heart, Mouse kidney; Cellular Location: Cell membrane, Cytoplasm, Endoplasmic reticulum membrane, Single-pass type I membrane protein, cytoskeleton.

STIM1 (stromal interacting molecule; also known as GOK) acts as a sensor of calcium depletion within the endoplasmic reticulum and transduces the signal to Orai1, the presumptive CRAC channel at the plasma membrane. Following decrease of luminal calcium concentration, STIM1 oligomerizes and induces Orai1 to enable entry of extracellular calcium into the cytoplasm. However, the precise mechanism of STIM1-Orai1 interaction has not been elucidated yet. Many questions also remain to be solved around STIM1 functional distribution. It turns out that STIM1 associates with growing ends of microtubules and is involved in endoplasmic reticulum tubule extension.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STIM1
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene STIM1
Gene Accession No. P70302, P84903, Q13586
Gene Alias D11S4896E; GOK; IMD10; RGD:1306831}; SIM; Stim1; stim1 {ECO:0000312; STIM1L; STRMK; Stromal interaction molecule 1; stromal interaction molecule 1 precursor; TAM; TAM1
Gene Symbols STIM1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 530-685 of human STIM1 (NP_0031472).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20866, 361618, 6786
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.