Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STIM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA146217
Description
The target sequence has 93% sequence homology with porcine and 79% sequence homology with xenopus. Suggested positive control: antigen standard for STIM1 (transient overexpression lysate), HeLa whole cell lysate.
STIM1 (stromal interacting molecule; also known as GOK) acts as a sensor of calcium depletion within the endoplasmic reticulum and transduces the signal to Orai1, the presumptive CRAC channel at the plasma membrane. Following decrease of luminal calcium concentration, STIM1 oligomerizes and induces Orai1 to enable entry of extracellular calcium into the cytoplasm. However, the precise mechanism of STIM1-Orai1 interaction has not been elucidated yet. Many questions also remain to be solved around STIM1 functional distribution. It turns out that STIM1 associates with growing ends of microtubules and is involved in endoplasmic reticulum tubule extension.
Specifications
STIM1 | |
Polyclonal | |
Unconjugated | |
STIM1 | |
D11S4896E; GOK; IMD10; RGD:1306831}; SIM; Stim1; stim1 {ECO:0000312; STIM1L; STRMK; Stromal interaction molecule 1; stromal interaction molecule 1 precursor; TAM; TAM1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20866, 361618, 534816, 608643, 6786 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 30% glycerol and 0.1% sodium azide | |
P70302, P84903, Q13586, Q58CP9 | |
STIM1 | |
Synthetic peptide made to a C-terminal region with residues 600-685 of the human protein.[Uniprot#Q13586]. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Canine, Bovine | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction