Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ STIM2 Polyclonal Antibody
STIM2 Polyclonal Antibody
Supplier: Thermo Scientific™ PA5109329
Description
STIM2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Specifications
STIM2 | |
Polyclonal | |
Unconjugated | |
STIM2 | |
Kiaa1482; OTTHUMP00000123457; OTTHUMP00000123458; OTTHUMP00000216198; Stim2; Stromal interaction molecule 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
57620 | |
-20°C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9P246 | |
STIM2 | |
Recombinant protein within Human STIM2 aa 1-140. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction