Learn More
Invitrogen™ STIM2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA517146
Catalog No. PIPA517146
Only null left
Add to Cart
Description
Description
It is not recommended to aliquot this antibody.
In T lymphocytes, the sole pathway for Ca++ entry following antigen-receptor binding is through store-operated Ca++-release-activated Ca++ (CRAC) channels. These channels are made up of the pore-forming subunit ORAI1 and the stromal interaction molecule 1 (STIM1), a protein that functions as a Ca++ sensor and activates the CRAC channels, migrating to the plasma membrane from endoplasmic reticulum (ER)-like sites which act as the Ca++ store. A related molecule, STIM2, acts to inhibit the STIM1-mediated store-operated Ca++ entry, and can form complexes with STIM1, suggesting they may play a coordinated role in controlling Ca++ entry. At least three isoforms of STIM2 are known to exist.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
STIM2
Polyclonal
0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
P83093, Q9P246
STIM2
Synthetic peptide corresponding to the sequence of human STIM2.
100 μL
Primary
Human, Mouse, Rat, Monkey
Antibody
IgG
Western Blot, Immunocytochemistry
Unconjugated
STIM2
Kiaa1482; OTTHUMP00000123457; OTTHUMP00000123458; OTTHUMP00000216198; Stim2; Stromal interaction molecule 2
Rabbit
Antigen affinity chromatography
RUO
116873, 117087, 57620
-20°C
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.