Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ STING agonist C53
GREENER_CHOICE

Catalog No. 774110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-411-0 10 mg
77-415-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 77-411-0 Supplier Tocris Bioscience™ Supplier No. 7741/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Pan-polymorph STING agonist

STING agonist C53 is an activator of human STING (EC50 = 185 nM). It promotes the oligomerization and activation of human STING through a mechanism orthogonal to that of cGAMP. STING agonist C53 increases activity of STING variants in HEK293T cells (107%, 92% and 92% reporter activity against H232, R232 and HAQ, respectively). It induces phosphorylation of STING variants in human tumor cell lines and PBMCs. STING agonist C53 is active at human but not mouse STING. Orally bioavailable.

Specifications

Chemical Name or Material 1-[(2-Chloro-6-fluorophenyl)methyl]-2,3-dihydro-3,3-dimethyl-2-oxo-N-[(2,4,6-trifluorophenyl)methyl]-1 H-indole-6-carboxamide
CAS 2259624-71-8
Quantity 10 mg
Target STING Agonists
Molecular Formula C25H19ClF4N2O2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.