Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STING/TMEM173 Antibody (2922D), Novus Biologicals™

Rabbit Monoclonal Antibody
$213.00 - $494.50
Specifications
Antigen | STING/TMEM173 |
---|---|
Clone | 2922D |
Concentration | 1.0 mg/ml |
Dilution | Western Blot 1 to 2 μg/mL, Simple Western, Immunohistochemistry 0.5 μg/mL, Immunocytochemistry/Immunofluorescence 3 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Description
STING/TMEM173 Monoclonal antibody specifically detects STING/TMEM173 in Human, Mouse, Rat samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
STING/TMEM173 | |
1.0 mg/ml | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Immunology, Innate Immunity | |
PBS | |
340061 | |
IgG | |
Protein A or G purified |
2922D | |
Western Blot 1 to 2 μg/mL, Simple Western, Immunohistochemistry 0.5 μg/mL, Immunocytochemistry/Immunofluorescence 3 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, hSTING, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, SAVI, Stimulator of interferon genes protein, stimulator of interferon protein, sting, STING-beta, TMEM173, transmembrane protein 173 | |
Partial recombinant protein made to amino acids 215-379 of human TMEM173/STING (UniProt Q86WV6). | |
Primary | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title