Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STING/TMEM173 Antibody (7C7), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | STING/TMEM173 |
---|---|
Clone | 7C7 |
Dilution | Western Blot 1:500-1:5000, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
STING/TMEM173 Monoclonal antibody specifically detects STING/TMEM173 in Human samples. It is validated for Western Blot, ELISASpecifications
STING/TMEM173 | |
Western Blot 1:500-1:5000, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, hSTING, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, SAVI, Stimulator of interferon genes protein, stimulator of interferon protein, sting, STING-beta, TMEM173, transmembrane protein 173 | |
A synthesized peptide derived from Human STING/TMEM173 [UniProt Q86WV6] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
7C7 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Cancer, Immunology, Innate Immunity | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
340061 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title