Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STING/TMEM173 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 22 publications
$244.50 - $574.50
Specifications
Antigen | STING/TMEM173 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1 - 2 ug/ml, Flow Cytometry 1-5 ug/million cells, ELISA reported in scientific literature (PMID 34905508), Immunohistochemistry 1:100 - 1:300, Immunocytochemistry/Immunofluorescence 5 - 10 ug/ml, Immunohistochemistry-Paraffin 1:100 - 1:300, Immunohistochemistry-Frozen reported in scientific literature (PMID 33745949) |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224683SS
![]() |
Novus Biologicals
NBP224683SS |
0.025 mg |
Each for $244.50
|
|
|||||
NBP224683
![]() |
Novus Biologicals
NBP224683 |
0.1 mg |
Each for $574.50
|
|
|||||
Description
STING/TMEM173 Polyclonal specifically detects STING/TMEM173 in Human, Mouse, Canine, Primate, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
STING/TMEM173 | |
Western Blot 1 - 2 ug/ml, Flow Cytometry 1-5 ug/million cells, ELISA reported in scientific literature (PMID 34905508), Immunohistochemistry 1:100 - 1:300, Immunocytochemistry/Immunofluorescence 5 - 10 ug/ml, Immunohistochemistry-Paraffin 1:100 - 1:300, Immunohistochemistry-Frozen reported in scientific literature (PMID 33745949) | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Primate | |
Q86WV6 | |
340061 | |
Partial synthetic peptide made to an internal portion of human STING/TMEM173 (between amino acids 310-360) [UniProt Q86WV6] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1.0 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Rabbit | |
Cancer, Immunology, Innate Immunity | |
PBS with 0.02% Sodium Azide | |
anti-STING, endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, human STING, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, mouse STING, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, Stimulator of interferon genes protein, sting, STINGhSTING, TMEM173, transmembrane protein 173 | |
TMEM173 | |
IgG | |
Peptide affinity purified | |
The amino acid sequence used as immunogen is 100% homologous in human, rhesus monkey, chimpanzee, and dog, 94% homologous in mouse and cow, 88% homologous in rat, 83% homologous in opossum and zebrafish, and 72% homologous in Xenopus. Peptide blocking: The response seen in western blot is reduced by over 50% in the presence of immunizing peptide. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title