Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STING/TMEM173 Antibody (STING1/8129R), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320616100UG
Description
STING/TMEM173 Monoclonal antibody specifically detects STING/TMEM173 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
STING/TMEM173 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, hSTING, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, SAVI, Stimulator of interferon genes protein, stimulator of interferon protein, sting, STING-beta, TMEM173, transmembrane protein 173 | |
Recombinant fragment (around aa190-290) of human STING/TMEM173 protein (exact sequence is proprietary) | |
100 μg | |
Cancer, Immunology, Innate Immunity | |
340061 | |
Store at 4&drg;C. Do not freeze. | |
IgG κ |
Immunohistochemistry (Paraffin) | |
STING1/8129R | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction